siRNA Details
Gene Symbol: Defb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031810.1 | NP_113998.1 | 2 | 218 | ||
GenBank mRNA | AF068860.1 | - | 2 | 194 | ||
AF093536.1 | - | 2 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
