siRNA Details
Gene Symbol: Cd164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031812.1 | NP_114000.1 | 3 | 286 | ||
GenBank mRNA | AJ238574.1 | - | 3 | 286 | ||
BC062064.1 | - | 3 | 289 | 5599253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
