siRNA Details
Gene Symbol: Clic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031818.1 | NP_114006.1 | 3 | 244 | ||
GenBank mRNA | AF104119.1 | - | 3 | 244 | ||
EF397567.1 | - | 3 | 244 | |||
FQ222737.1 | - | 354 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
