siRNA Details
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270800.1 | NP_001257729.1 | 5, 6 | 576 | ||
NM_001270801.1 | NP_001257730.1 | 5, 6 | 585 | |||
NM_031830.2 | NP_114018.1 | 5, 6 | 576 | |||
XM_006246952.1 | XP_006247014.1 | 6, 7 | 1197 | |||
XM_008768119.1 | XP_008766341.1 | 4, 5 | 563 | |||
GenBank mRNA | AF023302.1 | - | 5, 6 | 555 | ||
AF023303.1 | - | 5, 6 | 555 | |||
AF023304.1 | - | 4, 5 | 473 | |||
BC083550.1 | - | 4, 5 | 452 | 7133999 | ||
CB581049.1 | - | 5, 6 | 576 | |||
U64999.1 | - | 2, 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
