siRNA Details
Gene Symbol: Nme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031833.2 | NP_114021.2 | 5 | 712 | ||
GenBank mRNA | BC086599.1 | - | 5 | 712 | 7312852 | |
FQ217120.1 | - | 4 | 613 | |||
FQ217563.1 | - | 5 | 599 | |||
FQ218179.1 | - | 4 | 630 | |||
FQ222529.1 | - | 5 | 597 | |||
M55331.1 | - | 4 | 569 | |||
M91597.1 | - | 5 | 603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
