siRNA Details
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110333.2 | NP_001103803.1 | 6 | 1499 | ||
NM_001110335.1 | NP_001103805.1 | 4 | 626 | |||
NM_001110336.1 | NP_001103806.1 | 3 | 554 | |||
NM_001287107.1 | NP_001274036.1 | 7 | 1571 | |||
NM_001287108.1 | NP_001274037.1 | 6 | 1499 | |||
NM_001287113.1 | NP_001274042.1 | 5 | 1450 | |||
NM_031836.3 | NP_114024.2 | 7 | 1571 | |||
GenBank mRNA | AF062644.1 | - | 4 | 222 | ||
AF080594.1 | - | 2, 3 | 245 | |||
AF215725.1 | - | 7 | 557 | |||
AF260425.1 | - | 3 | 153 | |||
AF261751.1 | - | 4 | 226 | |||
AY033504.1 | - | 5 | 447 | |||
AY033506.1 | - | 6 | 496 | |||
AY033508.1 | - | 7 | 568 | |||
AY702972.1 | - | 6 | 499 | |||
BC168708.1 | - | 6 | 1375 | 7458665 | ||
CK357826.1 | - | 4 | 626 | |||
CK358638.1 | - | 1 | 66 | |||
CK364938.1 | - | 4 | 416 | |||
EU040284.1 | - | 3 | 144 | |||
EU040285.1 | - | 4 | 216 | |||
GQ423618.1 | - | 5 | 407 | |||
M32167.1 | - | 6 | 489 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001110266, NM_001025250, NM_009505, NM_001110267, BC061468, and others.
Gene Ontology Categories:
