siRNA Details
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110333.2 | NP_001103803.1 | 2, 3 | 1129 | ||
NM_001110334.2 | NP_001103804.1 | 2, 3 | 1129 | |||
NM_001287107.1 | NP_001274036.1 | 2, 3 | 1129 | |||
NM_001287108.1 | NP_001274037.1 | 2, 3 | 1129 | |||
NM_001287110.1 | NP_001274039.1 | 2, 3 | 1129 | |||
NM_001287111.1 | NP_001274040.1 | 2, 3 | 1129 | |||
NM_001287112.1 | NP_001274041.1 | 2, 3 | 1129 | |||
NM_031836.3 | NP_114024.2 | 2, 3 | 1129 | |||
GenBank mRNA | AF080594.1 | - | 1, 2 | 37 | ||
AF215725.1 | - | 2, 3 | 115 | |||
AF215726.1 | - | 2, 3 | 115 | |||
AF222779.1 | - | 2, 3 | 115 | |||
AY033506.1 | - | 2, 3 | 126 | |||
AY033507.1 | - | 2, 3 | 126 | |||
AY033508.1 | - | 2, 3 | 126 | |||
AY702972.1 | - | 2, 3 | 115 | |||
BC168708.1 | - | 2, 3 | 1005 | 7458665 | ||
CB810370.1 | - | 367 | ||||
GQ423618.1 | - | 1, 2 | 37 | |||
M32167.1 | - | 2, 3 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
