siRNA Details
Gene Symbol: Rps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031838.3 | NP_114026.3 | 3 | 316 | ||
GenBank mRNA | BC098726.1 | - | 3 | 305 | 7378399 | |
CB324420.1 | - | 4 | 335 | |||
CD372915.1 | - | 3 | 316 | |||
FQ209478.1 | - | 4 | 327 | |||
FQ210378.1 | - | 4 | 328 | |||
FQ213061.1 | - | 4 | 328 | |||
FQ213116.1 | - | 4 | 325 | |||
FQ214025.1 | - | 4 | 328 | |||
FQ215811.1 | - | 4 | 330 | |||
FQ216756.1 | - | 4 | 328 | |||
FQ218484.1 | - | 4 | 328 | |||
FQ218550.1 | - | 4 | 329 | |||
FQ218641.1 | - | 4 | 327 | |||
FQ218643.1 | - | 4 | 331 | |||
FQ219176.1 | - | 4 | 327 | |||
FQ219789.1 | - | 4 | 328 | |||
FQ219844.1 | - | 4 | 330 | |||
FQ219893.1 | - | 4 | 327 | |||
FQ220315.1 | - | 4 | 328 | |||
FQ220515.1 | - | 4 | 328 | |||
FQ220642.1 | - | 4 | 328 | |||
FQ220831.1 | - | 4 | 327 | |||
FQ220841.1 | - | 4 | 328 | |||
FQ220885.1 | - | 4 | 328 | |||
FQ220902.1 | - | 4 | 328 | |||
FQ221771.1 | - | 4 | 328 | |||
FQ221977.1 | - | 4 | 327 | |||
FQ222558.1 | - | 4 | 328 | |||
FQ222607.1 | - | 4 | 327 | |||
FQ222729.1 | - | 4 | 328 | |||
FQ222900.1 | - | 4 | 328 | |||
FQ223242.1 | - | 4 | 328 | |||
FQ225100.1 | - | 5 | 201 | |||
FQ225937.1 | - | 4 | 325 | |||
FQ226404.1 | - | 4 | 328 | |||
FQ226432.1 | - | 4 | 330 | |||
FQ226962.1 | - | 4 | 328 | |||
FQ228202.1 | - | 4 | 328 | |||
FQ229347.1 | - | 4 | 328 | |||
FQ229368.1 | - | 4 | 328 | |||
FQ229383.1 | - | 4 | 326 | |||
FQ229400.1 | - | 4 | 327 | |||
FQ229467.1 | - | 4 | 330 | |||
FQ229773.1 | - | 3 | 325 | |||
FQ229778.1 | - | 4 | 328 | |||
FQ230085.1 | - | 4 | 327 | |||
FQ230109.1 | - | 4 | 330 | |||
FQ230137.1 | - | 4 | 328 | |||
FQ230697.1 | - | 4 | 325 | |||
FQ231866.1 | - | 4 | 328 | |||
FQ233653.1 | - | 4 | 328 | |||
FQ234367.1 | - | 4 | 328 | |||
FQ234813.1 | - | 4 | 328 | |||
FQ235141.1 | - | 4 | 330 | |||
U92696.1 | - | 4 | 209 | |||
U92697.1 | - | 4 | 232 | |||
U92698.1 | - | 4 | 235 | |||
U92699.1 | - | 4 | 208 | |||
U92700.1 | - | 4 | 181 | |||
X57432.1 | - | 4 | 326 |
Gene Symbol: LOC100125366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC060584.1 | - | 2 | 519 | 6886957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
