siRNA Details
Gene Symbol: Ptms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031975.2 | NP_114181.1 | 5 | 787 | ||
GenBank mRNA | BC167753.1 | - | 5 | 777 | 7371571 | |
X16481.1 | - | 5 | 570 |

More Information
Other Species Targeted

Gene Ontology Categories:
