siRNA Details
Gene Symbol: Ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031979.3 | NP_114185.3 | 2, 3 | 478 | ||
GenBank mRNA | BC105778.1 | - | 2, 3 | 473 | 7932542 | |
D28557.1 | - | 2, 3 | 342 | |||
FQ226232.1 | - | 1, 2 | 27 | |||
U22893.1 | - | 2, 3 | 481 |

More Information
Other Species Targeted

Gene Ontology Categories:
