siRNA Details
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053341.1 | NP_445793.1 | 1, 2 | 345 | ||
XM_006255311.2 | XP_006255373.1 | 1, 2 | 2610 | |||
XM_006255312.2 | XP_006255374.1 | 2, 3 | 1604 | |||
XM_008772420.1 | XP_008770642.1 | 2, 3 | 1534 | |||
GenBank mRNA | AF032120.1 | - | 1, 2 | 345 | ||
AF089817.1 | - | 2, 3 | 363 | |||
BC070505.1 | - | 2, 3 | 388 | 7097697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
