siRNA Details
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053341.1 | NP_445793.1 | 1, 2 | 351 | ||
XM_006255311.2 | XP_006255373.1 | 1, 2 | 2616 | |||
XM_006255312.2 | XP_006255374.1 | 2, 3 | 1610 | |||
XM_008772420.1 | XP_008770642.1 | 2, 3 | 1540 | |||
GenBank mRNA | AF032120.1 | - | 1, 2 | 351 | ||
AF089817.1 | - | 2, 3 | 369 | |||
BC061964.1 | - | 2 | 1065 | |||
BC070505.1 | - | 2, 3 | 394 | 7097697 |
