siRNA Details
Gene Symbol: Nr5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191099.1 | NP_001178028.1 | 5 | 1036 | ||
XM_006234106.2 | XP_006234168.1 | 3 | 2047 |

More Information
Other Species Targeted

Gene Ontology Categories:
