siRNA Details
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053345.1 | NP_445797.1 | 3 | 211 | ||
XM_006243380.2 | XP_006243442.1 | 4 | 361 | |||
GenBank mRNA | AF000944.1 | - | 3 | 211 | ||
BC168727.1 | - | 5 | 395 | 6888477 | ||
FQ209546.1 | - | 366 | ||||
FQ212213.1 | - | 4 | 346 | |||
FQ221947.1 | - | 4 | 352 | |||
FQ228685.1 | - | 4 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
