siRNA Details
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031985.1 | NP_114191.1 | 7 | 698 | ||
XM_006247111.2 | XP_006247173.1 | 7 | 835 | |||
XM_008768120.1 | XP_008766342.1 | 7 | 1099 | |||
XM_008768121.1 | XP_008766343.1 | 8 | 721 | |||
GenBank mRNA | BC089789.1 | - | 8 | 768 | ||
M58340.1 | - | 7 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
