siRNA Details
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032067.1 | NP_114456.1 | 6 | 1548 | ||
GenBank mRNA | BC089788.1 | - | 6 | 1602 | 7374343 | |
U28830.1 | - | 6 | 1552 | |||
U82623.1 | - | 6 | 1548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
