siRNA Details
Gene Symbol: Ptger4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032076.3 | NP_114465.3 | 1 | 823 | ||
XM_006231996.2 | XP_006232058.1 | 2 | 3392 | |||
GenBank mRNA | D28860.1 | - | 2 | 1003 | ||
U94709.1 | - | 1 | 823 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008965, NM_001136079, BC011193, D13458, AK156428, and others.
Set Membership: |
![]() |
Gene Ontology Categories:
