siRNA Details
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032085.1 | NP_114474.1 | 48 | 3820 | ||
GenBank mRNA | AJ005395.1 | - | 10 | 1020 | ||
BC087039.1 | - | 48 | 3820 | 7097081 | ||
FQ214933.1 | - | 1 | 37 | |||
FQ215579.1 | - | 1 | 53 | |||
FQ215621.1 | - | 2 | 301 | |||
FQ228741.1 | - | 6 | 543 | |||
FQ229745.1 | - | 2 | 250 | |||
M21354.1 | - | 2 | 314 | |||
X70369.1 | - | 13 | 1236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
