siRNA Details
Gene Symbol: Sox11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053349.1 | NP_445801.1 | 1 | 1260 | ||
GenBank mRNA | AJ004858.1 | - | 1 | 1260 |

More Information
Other Species Targeted

Gene Ontology Categories:
