siRNA Details
Gene Symbol: Ackr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053352.1 | NP_445804.1 | 2 | 548 | ||
XM_006245479.2 | XP_006245541.1 | 3 | 742 | |||
XM_008767299.1 | XP_008765521.1 | 2 | 3867 | |||
XM_008767300.1 | XP_008765522.1 | 3 | 2828 | |||
GenBank mRNA | AF118816.1 | - | 2 | 548 | ||
AJ010828.1 | - | 2 | 543 |
