siRNA Details
Gene Symbol: Ikbkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053355.2 | NP_445807.2 | 4, 5 | 433 | ||
GenBank mRNA | AF115282.1 | - | 3, 4 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
