siRNA Details
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053357.2 | NP_445809.2 | 10 | 1708 | ||
XM_008766691.1 | XP_008764913.1 | 10 | 1791 | |||
GenBank mRNA | AF121265.1 | - | 10 | 1708 | ||
FQ222424.1 | - | 1901 | ||||
FQ224468.1 | - | 1906 |
