siRNA Details
Gene Symbol: Atox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053359.2 | NP_445811.1 | 3 | 201 | ||
GenBank mRNA | AF127137.1 | - | 3 | 137 | ||
AF177671.1 | - | 3 | 216 | |||
BC058458.1 | - | 3 | 225 | 6919265 | ||
FQ220104.1 | - | 3 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
