siRNA Details
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053369.1 | NP_445821.1 | 14 | 1486 | ||
GenBank mRNA | AF149284.1 | - | 14 | 1486 | ||
FQ221525.1 | - | 1237 | ||||
FQ226283.1 | - | 5 | 625 | |||
FQ227003.1 | - | 2 | 231 | |||
FQ232404.1 | - | 2 | 230 | |||
FQ233045.1 | - | 10 | 1118 | |||
U09228.1 | - | 8 | 893 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001083967, NM_013685, U16321, AK081012, AK028748, and others.
Gene Ontology Categories:
