siRNA Details
Gene Symbol: Timm10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053371.2 | NP_445823.1 | 67 | |||
GenBank mRNA | AF061242.1 | - | 2 | 32 | ||
AF150106.1 | - | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
