siRNA Details
Gene Symbol: Slpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053372.1 | NP_445824.1 | 1, 2 | 94 | ||
GenBank mRNA | AF151982.1 | - | 1, 2 | 94 | ||
AF178426.1 | - | 1, 2 | 90 | |||
AF421377.1 | - | 1, 2 | 104 |

More Information
Other Species Targeted

Gene Ontology Categories:
