siRNA Details
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053374.2 | NP_445826.2 | 3 | 370 | ||
XM_008759805.1 | XP_008758027.1 | 3 | 2528 | |||
GenBank mRNA | AF154569.1 | - | 2 | 181 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053374.2 | NP_445826.2 | 3 | 370 | ||
XM_008759805.1 | XP_008758027.1 | 3 | 2528 | |||
GenBank mRNA | AF154569.1 | - | 2 | 181 |