siRNA Details
Gene Symbol: Kif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053377.1 | NP_445829.1 | 4 | 560 | ||
XM_006246347.2 | XP_006246409.1 | 4 | 598 | |||
XM_006246351.2 | XP_006246413.1 | 4 | 597 | |||
XM_008767716.1 | XP_008765938.1 | 4 | 748 | |||
XM_008767717.1 | XP_008765939.1 | 4 | 597 | |||
GenBank mRNA | AM180763.1 | - | 2 | 152 | ||
CV116151.1 | - | 2 | 35 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
