siRNA Details
Gene Symbol: Atp1b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053381.1 | NP_445833.1 | 4 | 563 | ||
XM_006257493.2 | XP_006257555.1 | 4 | 854 | |||
XM_006257494.2 | XP_006257556.1 | 4 | 2288 | |||
XM_006257495.2 | XP_006257557.1 | 4 | 2273 | |||
GenBank mRNA | AF158385.2 | - | 4 | 563 | ||
AF158386.2 | - | 4 | 551 |

More Information
Other Species Targeted

Gene Ontology Categories:
