siRNA Details
Gene Symbol: Gjb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053388.1 | NP_445840.1 | 3 | 834 | ||
XM_006252079.2 | XP_006252141.1 | 3 | 2160 | |||
XM_006252080.2 | XP_006252142.1 | 3 | 1736 | |||
XM_008770781.1 | XP_008769003.1 | 3 | 948 | |||
XM_008770782.1 | XP_008769004.1 | 3 | 1258 | |||
XM_008770783.1 | XP_008769005.1 | 3 | 839 | |||
XM_008770784.1 | XP_008769006.1 | 3 | 945 | |||
GenBank mRNA | AF170284.1 | - | 3 | 834 | ||
BC078753.1 | - | 3 | 831 | 7132296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
