siRNA Details
Gene Symbol: Hs3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053391.1 | NP_445843.1 | 1 | 856 | ||
GenBank mRNA | AF177430.1 | - | 1 | 856 |

More Information
Other Species Targeted

Gene Ontology Categories:
