siRNA Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053420.3 | NP_445872.1 | 4, 5 | 497 | ||
GenBank mRNA | AF243515.1 | - | 4, 5 | 461 | ||
AY095482.1 | - | 4, 5 | 461 | |||
BC070958.1 | - | 4, 5 | 455 | 7105707 | ||
BC081690.1 | - | 4, 5 | 435 | 7190121 | ||
CO403082.1 | - | 4, 5 | 533 | |||
FQ213448.1 | - | 3, 4 | 258 | |||
FQ215108.1 | - | 4, 5 | 467 | |||
FQ215127.1 | - | 4, 5 | 467 | |||
FQ215203.1 | - | 4, 5 | 488 | |||
FQ216745.1 | - | 4, 5 | 488 | |||
FQ216768.1 | - | 1, 2 | 74 | |||
FQ216807.1 | - | 4, 5 | 488 | |||
FQ220853.1 | - | 4, 5 | 466 | |||
FQ221849.1 | - | 4, 5 | 488 | |||
KC660101.1 | - | 3, 4 | 294 |

More Information
Other Species Targeted

Gene Ontology Categories:
