siRNA Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053420.3 | NP_445872.1 | 2 | 211 | ||
GenBank mRNA | AF243515.1 | - | 2 | 175 | ||
AY095482.1 | - | 2 | 175 | |||
BC070958.1 | - | 2 | 169 | 7105707 | ||
BC081690.1 | - | 2 | 149 | 7190121 | ||
CO403082.1 | - | 2 | 247 | |||
FQ215108.1 | - | 2 | 181 | |||
FQ215127.1 | - | 2 | 181 | |||
FQ215203.1 | - | 2 | 202 | |||
FQ216745.1 | - | 2 | 202 | |||
FQ220853.1 | - | 2 | 180 | |||
FQ221849.1 | - | 2 | 202 | |||
KC660101.1 | - | 2 | 90 |
