siRNA Details
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053424.1 | NP_445876.1 | 23, 24 | 3147 | ||
XM_006250789.2 | XP_006250851.1 | 23, 24 | 3607 | |||
GenBank mRNA | AF004017.1 | - | 20, 21 | 2985 | ||
AF027362.1 | - | 20, 21 | 3017 | |||
AF210250.1 | - | 23, 24 | 3154 | |||
AF254802.1 | - | 23, 24 | 3147 |
