siRNA Details
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053425.1 | NP_445877.1 | 7 | 643 | ||
GenBank mRNA | AF255305.1 | - | 7 | 643 | ||
BC158586.1 | - | 7 | 603 | 9099085 | ||
EU340033.1 | - | 6 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
