siRNA Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053427.1 | NP_445879.1 | 9 | 1612 | ||
XM_006229265.1 | XP_006229327.1 | 9 | 1141 | |||
GenBank mRNA | AF271235.1 | - | 9 | 1612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
