siRNA Details
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109898.1 | NP_001103368.1 | 7 | 811 | ||
NM_053431.3 | NP_445883.1 | 7 | 811 | |||
GenBank mRNA | AB044285.1 | - | 7 | 811 | ||
AF217799.1 | - | 7 | 815 | |||
AF285078.1 | - | 7 | 833 | |||
AY623665.1 | - | 7 | 813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
