siRNA Details
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF217799.1 | - | 5 | 570 | ||
AF285078.1 | - | 5 | 588 | |||
AY623665.1 | - | 5 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
