siRNA Details
Gene Symbol: Xrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053435.1 | NP_445887.1 | 1, 2 | 139 | ||
XM_006228481.2 | XP_006228543.2 | 6, 7 | 778 | |||
XM_008758991.1 | XP_008757213.1 | 4, 5 | 640 | |||
GenBank mRNA | AF290895.1 | - | 1, 2 | 139 | ||
BC070894.1 | - | 1, 2 | 116 | 7099372 |
