siRNA Details
Gene Symbol: Stmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053440.2 | NP_445892.1 | 4 | 463 | ||
GenBank mRNA | AF306458.1 | - | 4 | 561 | ||
BC087660.1 | - | 4 | 463 | 7313867 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025285, BC026538, AK149212, AK003659, AK013020.
Gene Ontology Categories:
