siRNA Details
Gene Symbol: Rgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053453.2 | NP_445905.1 | 4, 5 | 448 | ||
GenBank mRNA | AF279918.1 | - | 461 | |||
AF321837.1 | - | 4, 5 | 448 | |||
AY043246.1 | - | 472 | ||||
BC061969.1 | - | 469 | 5621945 | |||
FQ224353.1 | - | 479 | ||||
FQ228949.1 | - | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
