siRNA Details
Gene Symbol: Plcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053456.1 | NP_445908.1 | 2 | 2906 | ||
XM_008767141.1 | XP_008765363.1 | 1 | 6687 | |||
GenBank mRNA | D45920.1 | - | 2 | 2906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
