siRNA Details
Gene Symbol: Cldn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053457.2 | NP_445909.2 | 3 | 747 | ||
GenBank mRNA | AF324043.1 | - | 3 | 552 | ||
BC070927.1 | - | 3 | 747 | 7096642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
