siRNA Details
Gene Symbol: Rab9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053458.1 | NP_445910.1 | 3 | 388 | ||
GenBank mRNA | AF325692.1 | - | 3 | 388 | ||
BC070502.1 | - | 3 | 392 | 7096737 | ||
FQ212962.1 | - | 3 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
