siRNA Details
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053460.1 | NP_445912.1 | 9 | 842 | ||
GenBank mRNA | AF327562.1 | - | 9 | 842 | ||
AF373289.1 | - | 9 | 764 | |||
AF542094.1 | - | 9 | 812 | |||
AJ240083.2 | - | 9 | 884 | |||
BC072689.1 | - | 10 | 845 | 7105747 |

More Information
Other Species Targeted

Gene Ontology Categories:
