siRNA Details
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053463.1 | NP_445915.1 | 6 | 744 | ||
XM_006229163.1 | XP_006229225.1 | 7 | 864 | |||
XM_006229164.2 | XP_006229226.1 | 7 | 808 | |||
XM_006229165.1 | XP_006229227.1 | 7 | 861 | |||
GenBank mRNA | AF336828.1 | - | 6 | 744 | ||
BC100643.1 | - | 7 | 794 | 7930904 | ||
Z36277.1 | - | 7 | 806 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008749, NM_001163662, AK170717, AK209002, AK004886, and others.
Gene Ontology Categories:
