siRNA Details
Gene Symbol: Ppp1r9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053473.2 | NP_445925.1 | 1 | 943 | ||
XM_006236057.2 | XP_006236119.1 | 2 | 1054 | |||
XM_006236058.2 | XP_006236120.1 | 2 | 1094 | |||
XM_006236061.2 | XP_006236123.1 | 2 | 1054 | |||
XM_006236063.2 | XP_006236125.1 | 2 | 1054 | |||
XM_006236064.2 | XP_006236126.1 | 2 | 1054 | |||
XM_006236066.2 | XP_006236128.1 | 2 | 1051 | |||
XM_006236068.2 | XP_006236130.1 | 2 | 1050 | |||
GenBank mRNA | U72994.1 | - | 1 | 947 |
