siRNA Details
Gene Symbol: Pola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053479.1 | NP_445931.1 | 20 | 2326 | ||
GenBank mRNA | AJ011605.1 | - | 20 | 2326 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008892, D13543, D17384.
Gene Ontology Categories:
