siRNA Details
Gene Symbol: Pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053487.2 | NP_445939.1 | 2 | 185 | ||
XM_008759560.1 | XP_008757782.1 | 1 | 37 | |||
GenBank mRNA | AJ224120.1 | - | 2 | 217 |

More Information
Other Species Targeted

Gene Ontology Categories:
