siRNA Details
Gene Symbol: Nme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053507.1 | NP_445959.1 | 3 | 242 | ||
XM_008767598.1 | XP_008765820.1 | 3 | 354 | |||
GenBank mRNA | AY017337.1 | - | 3 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
